Learn More
World Precision Instrument Harmon Forceps, 4x5 teeth, 7.5cm
SDP
Supplier:  World Precision Instrument 504484
Encompass_Preferred
  • Harmon Forceps
  • 7.5cm (3") Long
  • 4x5 teeth, 2.5mm jaw width
  • Stainless steel
Catalog No. 50-253-9365