Learn More
Grainger Harness Quick Connector Nano-Lok
Supplier:  Grainger 16V475
Encompass_Preferred
The 3M DBI-SALA Harness Quick Connector 420 is a quick-connect harness connector designed for use with DBI-SALA ExoFit and Ultra-Lok harnesses. It allows for quick and easy connections and disconnections. This connector has a working load limit of 420 lbs and is made from zinc plated steel for durability. It has an opening gate with dual action locking tabs for security. This product meets OSHA and ANSI standards. The connector's size is 3.6 x 2.6 x 1.3 inches and it weighs 0.32 lbs.
Catalog No. 50-276-6672