Learn More
Medchemexpress LLC Harpagoside | 19210-12-9 | 494.49 | 20 MG
SDP
Supplier:  Medchemexpress LLC HYN039620MG
Encompass_Preferred
Harpagoside, derived from *Harpagophytum procumbens*, demonstrates anti-inflammatory, anti-cancer, and protective activities. It exhibits inhibitory effects on COX-1 and COX-2 activity, suppresses NO production, and inhibits HepG2 cell lipid polysaccharide. This compound also possesses anti-inflammatory and latent pain effects, protecting the body and showing a degenerative effect on β-oxidation (Aβ).
  • Exhibits anti-inflammatory, anti-cancer, and protective activities.
  • Inhibits COX-1 and COX-2 activity.
  • Suppresses NO production.
  • Inhibits HepG2 cell lipid polysaccharide.
  • Possesses anti-inflammatory and latent pain effects.
  • Protects the body and shows a degenerative effect on β-oxidation (Aβ).
Catalog No. 50-003-88697