Learn More
Medchemexpress LLC Harpagoside | 19210-12-9 | 494.49 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN03965MG
Encompass_Preferred
Harpagoside can be obtained from *Harpagophytum procumbens*. It exhibits anti-inflammatory, anti-cancer, and protective activities. Harpagoside inhibits COX-1 and COX-2 activity, suppresses NO production, and inhibits HepG2 cell lipid polysaccharide expression, leading to anti-inflammatory and latent pain effects. It also has a protective and degenerative effect on β-oxidation (Aβ).
  • Anti-inflammatory activity.
  • Anti-cancer activity.
  • Protective activity.
  • Inhibitory effect on COX-1 and COX-2.
  • Suppresses NO production.
  • Inhibits HepG2 cell lipid polysaccharide expression.
  • Potential neuroprotective effects against Aβ intoxication in an Alzheimer's disease model.
Catalog No. 50-003-89650