Learn More
TARGETMOL CHEMICALS INC Harpagoside 20MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T6S202320MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 1. Harpagoside has anti-inflammatory activity. 2. Harpagoside at 3 mM concentration, shows moderate inhibition of seed germination. 3. Harpagoside can inhibit lipopolysaccharide-induced mRNA levels and protein expression of cyclooxygenase-2 and inducible nitric oxide in HepG2 cells. Purity 100%
Catalog No. 50-287-8105