Learn More
Cayman Chemical HarpagosIde 50mg
SDP
Supplier:  Cayman Chemical 3896750 MG
Encompass_Preferred
An iridoid glycoside with diverse biological activities; inhibits LPS-induced increases in COX-2 and NO levels, as well as TNF mRNA expression in HepG2 cells from 50-200 µM; reduces synovial inflammation, joint destruction, and bone erosion in a mouse model of collagen-induced arthritis at 10 mg/kg; decreases substantia nigra pars compacta neuronal cell death and increases time spent on the rod in a rotarod test in a mouse model of MPTP-induced Parkinson’s disease
Catalog No. 50-291-7314