Learn More
Selleck Chemical LLC Harringtonine 1mg
SDP
Supplier:  Selleck Chemical LLC S90631MG
Encompass_Preferred
Harringtonine is a natural cephalotaxine alkaloid that inhibits protein biosynthesis. It also exhibits antiviral and anticancer activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3468