Learn More
Electron Microscopy Sciences Harris Uni-Core™ Sampling Tool 1.0
SDP
Supplier:  Electron Microscopy Sciences 6903610
Encompass_Preferred
The Harris Uni-Core is a disposable multi-purpose sampling tool for soft substrates. Made from polypropylene, including a hollow body with built-in tool steel razor sharp cutting tip, with a plunger for the expelling of the punched sample. Uni-Core comes individually wrapped and sterilized by ethylene oxide. Uni-Core can also be reusable by rinsing the tip with ethanol after each use and autoclaving for 20 minutes at 250 degree F/15 psi.
Catalog No. NC0711699