Learn More
Cayman Chemical HarrngtonIn 10mg
SDP
Supplier:  Cayman Chemical 1536110 MG
Encompass_Preferred
A natural alkaloid that inhibits protein synthesis at low micromolar concentrations; immobilizes ribosomes immediately after the initiation of translation
Catalog No. 50-296-9253