Learn More
ABclonal Technology HAS3 Rabbit pAb
SDP
Supplier:  ABclonal Technology A1349820UL
Encompass_Preferred
The protein encoded by this gene is involved in the synthesis of the unbranched glycosaminoglycan hyaluronan, or hyaluronic acid, which is a major constituent of the extracellular matrix. This gene is a member of the NODC/HAS gene family. Compared to the proteins encoded by other members of this gene family, this protein appears to be more of a regulator of hyaluronan synthesis. Alternative splicing results in multiple transcript variants.
Catalog No. 50-000-98531