Learn More
MSC HAZCO Corrugated Shipping Box, 5 in High, 1 ea
Supplier: MSC 39549761
This corrugated shipping box is designed for hazardous materials transport, offering a 95 lb capacity and durable Kraft construction. It features a single wall design for efficient handling and is suitable for various shipping applications.
- Made in USA
- Five in high
- One wall construction
- Kraft material
- Offers a 95 lb load capacity
- Rectangle container shape
- Brown color
- Printed with mandatory warning information
- Features 'This side up' arrows printed on two sides
- Used as refills for foam shipper kits
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.