Learn More
Medchemexpress LLC HB-EGF Mouse 500ug

Supplier: Medchemexpress LLC HYP7194500UG
Recombinant mouse HB-EGF (heparin-binding EGF-like growth factor) is supplied as a lyophilized, biologically active protein for in vitro research into wound healing, cardiac hypertrophy, and heart development. The product is characterized for purity and specific activity, and includes recommended formulation and storage guidance to help maintain stability and activity.
- Expressed in E. coli for recombinant production.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
- High specific activity with ED50 <1 ng/mL in murine BALB/c 3T3 cells.
- Lyophilized formulation for extended shelf life and easy reconstitution.
- Recommended storage and handling to preserve activity (aliquot and store at -20°C or -80°C).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.