Learn More
Medchemexpress LLC HB-EGF Mouse 5ug

Supplier: Medchemexpress LLC HYP71945UG
HB-EGF Protein, Mouse is a lyophilized recombinant heparin-binding EGF-like growth factor (mouse sequence) supplied for research use in cell-based assays and biochemical studies. The product is expressed in Escherichia coli, characterized by SDS-PAGE and activity assays, and is intended for investigations of wound healing, cardiac hypertrophy, and heart development.
- Recombinant mouse HB-EGF expressed in Escherichia coli.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Predicted molecular mass ~9.8 kDa; appears ~10 kDa on SDS-PAGE.
- Lyophilized formulation with stabilizers for storage and transport.
- High biological activity: ED50 < 1 ng/mL in murine Balb/c 3T3 cell assay.
- Available in small pack sizes appropriate for cell-based assays, including 5 μg.
- Store lyophilized at -20°C; aliquot and include carrier protein for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.