Learn More
INTACT GENOMICS INC HB101 Electrocompetent Cells 24x50μl
Supplier:  INTACT GENOMICS INC 127148
Encompass
Intact Genomics (ig) HB101 electrocompetent E. coli cells are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. E. coli HB101 is a K12 x B hybrid strain, containing the recA13 mutation that minimizes recombination and helps insert stability. In addition, it carries the hsdS20(rB-mB-) restriction minus genotype which prevents cleavage of cloned DNA by endogenous restriction enzymes. HB101 strain does not support Alpha-complementation for blue/white screening.Product Include and Storage: ig HB101 electrocompetent cells: -80 ºCpUC19 control DNA: -20 ºCRecovery medium: 4 ºC
Catalog No. 50-000-82948