Learn More
TARGETMOL CHEMICALS INC HBED 100MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T32046100MG
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HBED (CHELII) is an iron chelator and can be used in research on the treatment of chronic iron overload and acute iron poisoning. purity: 99%
Catalog No. 50-257-6977