Learn More
Medchemexpress LLC HBED 10mg | 35998-29-9 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11483710MG
Encompass_Preferred
HBED is a synthetic iron chelator used in research to bind and sequester iron; it is commonly used to study transfusional iron overload and iron-mediated pathways such as ferroptosis. The compound is supplied as a white to off-white solid, with molecular formula C20H24N2O6 and molecular weight 388.41 g/mol.
  • Potent iron chelator for in vitro and ex vivo studies.
  • Useful for modeling transfusional iron overload and related pathways.
  • White to off-white powder form suitable for dissolution in common solvents.
  • Stable as a powder at -20°C for extended storage.
  • Available in small milligram quantities for laboratory research.
Catalog No. 50-002-26405