Learn More
Medchemexpress LLC HBED-CC-tris(tert-butyl ester) TFA | 2097123-80-1 | C40H57F3N2O12 | 50 MG
Supplier: Medchemexpress LLC HY15355150MG
HBED-CC-tris(tert-butyl ester) TFA is a derivative of HBED-CC and a bifunctional chelator. It can be used in the research of radiolabeling and compounds for tumor-assisted diagnosis. When conjugated with PSMA and labeled with gallium isotopes, it can be used in the research on the auxiliary diagnosis of prostate cancer.
- Derivative of HBED-CC
- Bifunctional chelator
- Used in research of radiolabeling
- Used in research of compounds for tumor-assisted diagnosis
- Can be conjugated with PSMA
- Can be labeled with gallium isotopes
- Used in research on the auxiliary diagnosis of prostate cancer