Learn More
Promocell GmbH PromoCell HBEpC-c 500,000 Human Bronchial Epithelial Cells, cryopreserved
Supplier:  Promocell GmbH C12640
Encompass
Primary Human Bronchial Epithelial Cells (HBEpC) are isolated from the surface epithelium of human bronchi and stain positive for cytokeratin.The respiratory epithelia are responsible for the lubrication of the lungs, the maintenance of humidity, and the cleaning of the respiratory tract. They are an important target for drugs, toxins, and carcinogens. In addition, they are involved in many diseases such as cystic fibrosis. Consequently, HBEpC are useful for investigating the function and pathology of the respiratory system.Our HBEpC are now also available from Air-Liquid Interface pre-screened, HLA-typed, and COPD/Asthma donors.
Catalog No. 50-306-046