Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrazolo[1,5-a]pyrimidine, 7-(2-chloro-6-fluorophenyl)-5-(4-chlorophenyl)-1,5,6,7-tetrahydr | 957011-15-3 | 99.99% | 1 ML
SDP

Supplier:  Medchemexpress LLC HY12697010MM/1ML

Encompass_Preferred

HBF-0259 is a potent and selective inhibitor of hepatitis B virus (HBV) surface antigen (HBsAg) secretion, with an EC50 of 1.5 μM in HepG2.2.15 cells. It has no effect on HBV DNA synthesis.

  • Potent and selective inhibitor of HBsAg secretion
  • Has no effect on HBV DNA synthesis
  • For research use only
  • Available as a white to off-white solid
  • High purity of 99.99%
  • Soluble in DMSO at 100 mg/mL
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-41323


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.