Learn More
Cayman Chemical HBP08trIfluoroacetAT salt 10mg
SDP
Supplier:  Cayman Chemical 3696110 MG
Encompass_Preferred
A peptide inhibitor of the CXCL12-HMGB1 protein-protein interaction; binds to HMGB1 (Kd = 0.8 µM); inhibits HMGB1-induced inhibition of CXCL12-induced migration of 300-19 pre-B cells expressing human CXCR4
Catalog No. 50-291-5459