Learn More
Creative Biomart Recombinant HBsAg adw Antigen
Supplier:  Creative Biomart HBS075
Encompass_Preferred
HbsAg adw produced Pichia Pastoris, having a molecular weight of approximately 24.0 kDa as shown on SDS-PAGE.
Catalog No. 50-208-4453