Learn More
Medchemexpress LLC HY-17540 10mg , HBX 19818 CAS:1426944-49-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1754010MG
Encompass_Preferred
HY-17540 10mg HBX 19818 CAS: 1426944-49-1 HBX 19818 is a specific inhibitor of ubiquitin-specific protease 7 (USP7), with an IC50 of 28.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1004