Learn More
Apexbio Technology LLC HC 067047,10mg CAS# 883031-03-6
SDP
Supplier:  Apexbio Technology LLC B758810
Encompass_Preferred
HC067047 is a selective TRPV4 antagonist with IC50 of 48 nM, 133 nM and 17 nM for human, rat and mouse TRPV4, respectively[1]. In HEI-OC1 cells, HC067047 (1 µM) treatment for 24 hours can significantly reduce the mRNA expression in HEI-OC1 cells cultured in high glucose and reduce the expression of TRPV4 protein. HC067047 (1 µM) treatment for 48 hours can inhibit cell proliferation and promote apoptosis[2]. In WT mice with cystiti, HC067047 (0-50 mg/kg; intraperitoneal injection; for 30 minutes) treatment increases functional bladder capacity and reduces micturition frequency. HC067047 do not affect bladder function in Trpv4−/− mice[1].  Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0045