Learn More
Selleck Chemical LLC HC-067047-5mg
SDP
Supplier:  Selleck Chemical LLC S66375MG
Encompass_Preferred
HC-067047 is a selective TRPV4 channel antagonist with IC50 values of 48 nM,133 nM,and 17 nM for human, rat, and mouse TRPV4 orthologs,respectively.
Catalog No. 50-238-9766