Learn More
Medchemexpress LLC HC-070 | 1628291-95-1 | 99.7% | 475.32 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY112302100MG
Encompass_Preferred
HC-070 is a selective antagonist of TRPC4/TRPC5 channels, inhibiting hTRPC5 with an IC50 of 9.3 nM and hTRPC4 with an IC50 of 46 nM. It exhibits over 400-fold selectivity for human TRPC4/5 channels over other examined channels and weakly inhibits TRPC3 (IC50, 1 μM). It blocks various TRPC-mediated currents, including lanthanum-activated hTRPC5 and M2R-activated human TRPC1/TRPC4. In animal models, HC-070 has been shown to reduce CCK-4 evoked neuronal activity in the amygdala, decrease anxiety, and alleviate fear memory and marble burying behavior without affecting locomotor activity.
  • Selective TRPC4/TRPC5 antagonist
  • High selectivity for human TRPC4/5 channels
  • Modulates TRPC-mediated currents
  • Reduces neuronal activity
  • Decreases anxiety and fear memory
  • Reduces marble burying behavior
Catalog No. 50-004-38207