Learn More
Medchemexpress LLC HC-070 | 1628291-95-1 | 99.74% | 475.32 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11230210MG
Encompass_Preferred
HC-070 is a potent and selective antagonist of TRPC4 and TRPC5 channels, demonstrating IC50s of 9.3 nM for hTRPC5 and 46 nM for hTRPC4. It exhibits high selectivity, with at least 400-fold preference for human TRPC4 and TRPC5-containing channels over other examined channels. This compound also modulates TRPC1/TRPC4 and TRPC1/5 channels and reduces neuronal activity in amygdala slices.
  • Inhibits TRPC4/TRPC5 channels.
  • Shows high selectivity for TRPC4 and TRPC5-containing channels.
  • Modulates TRPC1/TRPC4 and TRPC1/5 channels.
  • Reduces neuronal activity in amygdala slices.
  • Weakly inhibits TRPC3.
  • Inhibits lanthanum-activated TRPC5-mediated currents.
  • Blocks M2R-activated TRPC1/TRPC4 channels.
  • Inhibits human TRPC5 currents activated via muscarinic type 1 (M1R).
  • Suppresses hTRPC4 currents via M2R.
Catalog No. 50-004-38265