Learn More
ATCC HC11; Mammary Epithelium; Mouse (Mus musculus)
Supplier:  ATCC CRL3062
Encompass
HC11 Mammary Epithelium is an epithelial cell line isolated in 1999 from the mammary glands of a mouse. It was deposited by Harvard Medical School Department of Genetics can be used as an in vitro model to study mammary epithelial cell proliferation, signal transduction, and differentiation.
Catalog No. 50-238-4863
 Restricted Item