Learn More
ATCC Primary Human Coronary Artery Smooth Muscle Cells (HCASMC) - Normal, Cryopreserved Primary Cells
Supplier: ATCC PCS100021
Primary human coronary artery smooth muscle cells (HCASMC) are normal, primary smooth muscle cells isolated from human coronary artery tissue. These cryopreserved cells are used for in vitro studies of vascular biology, smooth muscle function, proliferation, signaling, and disease modeling.
- Species: human (Homo sapiens).
- Cell type: coronary artery smooth muscle cell.
- Cell status: normal, primary cells.
- Format: cryopreserved vials for culture.
- Recommended culture: vascular cell basal medium with smooth muscle growth supplements.
- Common applications: vascular biology, pharmacology, gene expression, and in vitro contractility studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.