Learn More
Medchemexpress LLC HCC-1/CCL14 Human 500ug
SDP
Supplier:  Medchemexpress LLC HYP7195500UG
Encompass_Preferred
Recombinant human HCC-1/CCL14 (residues T22-N93) is a CC chemokine produced in Escherichia coli for use in cell biology and immunology research. It is used to study chemotactic responses and signaling pathways in monocytes, eosinophils, and T lymphocytes, and is supplied in research-scale quantities suitable for in vitro assays.
  • Recombinant human chemokine corresponding to residues T22-N93.
  • Expressed in Escherichia coli for consistent production.
  • Enables in vitro chemotaxis and signaling assays.
  • Useful for studies of monocyte, eosinophil, and T-lymphocyte responses.
  • Provided in research-scale quantities suitable for assay development.
Catalog No. 50-001-95958