Learn More
ATCC HCC1937; Breast Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2336
Encompass
HCC1937 were isolated from a primary ductal carcinoma derived from a 23-year-old, White female with stage IIB breast cancer. This cell line was initiated in 1995 and took 11.5 months to establish.
Catalog No. 50-238-4521