Learn More
ATCC HCC38; Breast Ductal Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CRL2314
Encompass
HCC38 cells are epithelial cells isolated from the mammary gland of a 50-year-old, White female with a prior history of leiomyosarcoma; her mother died of breast cancer. It can be used in breast cancer research. This cell line was initiated on 4/27/92 and took 32 months to establish.
Catalog No. 50-238-1966