Learn More
Medchemexpress LLC HY-P1014 1mg Medchemexpress, HCGRP-(8-37) CAS:119911-68-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP10141MG
Encompass_Preferred
Medchemexpress, HY-P1014 1mg HCGRP-(8-37) CAS:119911-68-1 HCGRP-(8-37) is a human calcitonin gene-related peptide (hCGRP) fragment and also an antagonist of CGRP receptor . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4129