Learn More
Medchemexpress LLC HCH6-1 | 1435265-06-7 | 99.2% | 469.53 | C28H27N3O4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10128310MM 1ML
Encompass_Preferred
HCH6-1 is a potent, competitive dipeptide antagonist of the formyl peptide receptor 1 (FPR1) used in pharmacology research to study FPR1-mediated neutrophil responses and inflammatory processes. It is supplied in research-grade formats suitable for in vitro and in vivo studies.
  • Potent FPR1 antagonist that inhibits chemotaxis and superoxide generation.
  • Suitable for research into FPR1-related inflammatory diseases.
  • Available as 10 mM solution in DMSO and multiple solid pack sizes for flexibility.
  • High chemical purity (>99%) suitable for sensitive assays.
  • Characterized molecular weight and formula for reliable identification.
Catalog No. 50-002-12775