Learn More
Medchemexpress LLC Hch6-1 | 1435265-06-7 | 99.2% | 469.53 | C28H27N3O4 | 50 MG

Supplier: Medchemexpress LLC HY10128350MG
HCH6-1 is a potent, competitive dipeptide antagonist of formyl peptide receptor 1 (FPR1) used for research into neutrophil activation and inflammatory lung disease. It inhibits chemotaxis, superoxide anion generation, and elastase release, and has shown protective effects in acute lung injury models.
- Potent FPR1 antagonist effective in neutrophil assays.
- Inhibits chemotaxis, superoxide production, and elastase release.
- Molecular formula C28H27N3O4; molecular weight 469.53.
- CAS number 1435265-06-7.
- Purity 99.16% (reported).
- Solid, white to off-white appearance.
- Soluble in DMSO (250 mg/mL); in vivo formulations ≥ 2.08 mg/mL.
- Recommended storage: powder -20°C for long term or 4°C for shorter term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.