Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMarKa Chemical Company Hydrochloric Acid 6N Low Si 1L

Supplier:  LaMarKa Chemical Company 0000069616750101

Encompass_Preferred

Hydrochloric Acid 6N Si Free-Bottle Liter Plastic

Catalog No. 50-440-0200


May include imposed supplier surcharges.
 Restricted Item
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.