Learn More
ATCC HCM-CSHL-0084-C25 (ATCC PDM-34) Patient-derived Pancreatic Organoid - Pancreatic Ductal Adenocarcinoma, for research use
Supplier:  ATCC HCM CSHL0084C25
Encompass
HCM-CSHL-0084-C25 (ATCC PDM-34) is a patient-derived organoid model isolated from primary pancreatic ductal adenocarcinoma tissue. It is intended for laboratory research use and is suitable for basic cancer biology studies and pharmacological screening applications.
  • Patient-derived organoid model isolated from primary pancreatic ductal adenocarcinoma
  • Suitable for basic cancer biology and pharmacological screening applications
  • Organoid morphology supports three-dimensional cell culture studies
  • Biosafety level 1 and intended for research use only
  • Not for therapeutic, diagnostic, or human or animal consumption use
Catalog No. NC3815856