Learn More
ATCC ATCC PDM-514 (HCM-WCMC-0673-C56-B) Human Ovarian Organoid Cell Model - 3D Culture, Cancer Research, BSL-1
Supplier:  ATCC PDM514
Encompass
HCM-WCMC-0673-C56-B (PDM-514) is a patient-derived organoid model established from primary ovarian cancer tissue and developed through the Human Cancer Models Initiative for use in cancer research and compound screening. The product is supplied frozen and supports embedded 3D culture workflows for assay development and pharmacological testing.
  • Patient-derived from primary ovarian tumor tissue.
  • Supplied frozen with ≥ 1.0 x 10^6 cells per vial.
  • Designed for embedded 3D organoid culture.
  • Suitable for cancer research, screening, and basic research applications.
  • Maintain at 37 °C in 95% air, 5% CO2 and renew media every 2-3 days.
  • Store in the vapor phase of liquid nitrogen for long-term preservation.
Catalog No. NC2855748