Learn More
ATCC HCM-CSHL-0060-C18; Cancer Model Primary; Adenocarcinoma Sigmoid Colon; Human (Homo sapiens)
Supplier:  ATCC PDM4
Encompass
A patient-derived next-generation cancer model generated by the Human Cancer Models Initiative (HCMI). HCM-CSHL-0060-C18 (ATCC No. PDM-4) was isolated from primary adenocarcinoma of large intestine tissue. This tumor-derived model can be used in basic research and pharmacological screening applications. Data for the parental tumor and the tumor-derived organoid models are available at the GDC. Additional molecular characterizations may be available at the GDC. Additional controlled data may be available via dbGaP.
Catalog No. 50-238-4273