Learn More
Sino Biological HCoV-HKU1 Spike PAb 100ul (Sino Biological)
Supplier:  Sino Biological 40021T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant HCoV-HKU1 (isolate N1) Spike/S1 ( Catalog#40021-V08H; YP_173238.1; Met1-Arg760). HCoV-HKU1 (isolate N1) Spike/S1 specific IgG was purified by HCoV-HKU1 (isolate N1) Spike/S1 affinity chromatography.
Catalog No. 50-237-0537