Learn More
Sino Biological Human coronavirus (HCoV-NL63) Spike S1+S2 Protein (ECD, His Tag), Biotinylated
Supplier:  Sino Biological 40604V08BB20
Encompass_Preferred
A DNA sequence encoding the human coronavirus (HCoV-NL63) spike protein (S1+S2 ECD) (APF29071.1) (Met1-Pro1296) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. NC2072268