Learn More
AdipoGen HCPI
SDP
Supplier:  AdipoGen CDXH0003M250
Encompass_Preferred
Chemical. CAS 151589-38-7. Formula C22H18N4O. MW 354.4. Synthetic. 2-4-Hydrazinocarbonylphenyl-4,5-diphenylimidazole HCPI as a lophine derivative is a versatile fluorescent derivatization reagent for fatty acids. HCPI has a hydrazinocarbonyl group which reacts with fatty acids resulting in intensely fluorescent derivatives.
Catalog No. 50-256-0810