Learn More
Eurofins Discoverx PathHunter HCRTR2 Total GPCR Internalization U2OS Cell Line, 2 vials. (93-0730C3)
Supplier:  Eurofins Discoverx 930730C3
Encompass
The PathHunter HCRTR2 Total GPCR internalization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This cell line uses EFC technology to measure ligand-induced internalization of HCRTR2 via detection of localization to the early endosome. The included cell line overexpresses ProLink-tagged HCRTR2 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the HCRTR2 receptor to the early endosome leads to production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-1203
Add to Cart
Request a Quote