Learn More
ATCC HCT 116; Colon Carcinoma; Human (Homo sapiens)
Supplier:  ATCC CCL247
Encompass
HCT 116 cell line was isolated from the colon of an adult male with colon cancer. It has a mutation in codon 13 of the ras proto-oncogene and can be used as a positive control for PCR assays of mutation in this codon.
Catalog No. 50-238-3906
 Restricted Item