Learn More
Medchemexpress LLC HCV-IN-31 | 1998705-62-6 | 99.7% | 312.30 | 1 ML

Supplier: Medchemexpress LLC HY13830510MM/1ML
HCV-IN-31 is a potent HCV inhibitor. It has an EC50/EC95 of 15.7 μM for HCV replicon, making it suitable for research applications related to hepatitis C virus inhibition.
- Functions as an HCV inhibitor
- Exhibits an EC50/EC95 of 15.7 μM for HCV replicon
- Solid appearance
- Color ranges from white to off-white
- Purity of 99.7%
- Molecular weight of 312.30
- Intended for research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.