Learn More
Medchemexpress LLC HCV-IN-31 | 1998705-62-6 | 99.7% | 312.30 | 50 MG

Supplier: Medchemexpress LLC HY13830550MG
HCV-IN-31 is an HCV inhibitor, also known as compound 4. It exhibits an EC50/EC95 of 15.7 μM for HCV replicon.
- Solid appearance: White to off-white
- Purity: 99.69%
- Molecular weight: 312.30
- Solid storage: 4°C, protected from light
- Storage in solvent: -80°C for 6 months; -20°C for 1 month, protected from light
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.