Learn More
MSC Baldwin Filters Fuel Element, Bail Handle, 9.8 Nominal Micron, 1 ea
Supplier:  MSC 55868848
Encompass
The PF809 HD fuel element is designed for heavy-duty filtration applications in various industrial and agricultural equipment. This fuel element ensures engine efficiency by maintaining a system free of particulate and water contamination.
  • Features a bail handle for ease of installation
  • Nominal micron rating of 9.8
  • Includes G104 gasket
  • Equipped with an attached grommet
  • Compatible with diverse equipment including Allis Chalmers, Case, Caterpillar, and Massey Ferguson
  • Outside diameter of 4 7/32 in. and inside diameter of 1/2 in.
  • Overall length of 4 7/8 in.
Catalog No. 50-005-08302