Learn More
Enzo Life Sciences FLUOR DE LYS-HDAC8 deacetylase substrate (0.5 µmol)
SDP
Supplier:  Enzo Life Sciences BMLKI1780005
Encompass_Preferred
A fluorogenic, diacetylated peptide substrate for HDAC8 (histone deacetylase-8). Formulation: Supplied as a 5 mM solution (100 µl) in HDAC Assay Buffer.
Catalog No. 50-210-0528