Learn More
Epigentek Inc Epigenase HDAC Activity/Inhibition Direct Assay Kit (Colorimetric) (48 assays)
Supplier: Epigentek Inc P403448
The Epigenase HDAC Activity/Inhibition Direct Assay Kit is a complete set of optimized buffers and reagents for measuring the activity or inhibition of total HDAC enzymes in nuclear extracts or purified HDAC isoforms (HDACs 1-11) from a broad range of species such as mammals, plants, fungi, and bacteria, and in a variety of forms including, but not limited to, cultured cells and fresh and frozen tissues. The Epigenase HDAC Activity/Inhibition Direct Assay Kit is a further refinement of its predecessor, the EpiQuik HDAC Activity/Inhibition Assay Kit, a popular assay method for HDAC activity and inhibition. This latest method retains the simplicity, rapidness, high throughput capability, and non-radioactivity featured in the previous version.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.