Learn More
Medchemexpress LLC HDAC-IN-27 dihydrochloride | 3069831-25-7 | ≥99.0% | C20H24Cl2N4O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY142690A10MG
Encompass_Preferred
HDAC-IN-27 dihydrochloride is a potent, class I-selective histone deacetylase (HDAC) inhibitor provided as the dihydrochloride salt for research use. It exhibits low-nanomolar inhibition of HDAC1-3, shows both in vitro and in vivo antitumor activity, and promotes histone acetylation and apoptosis in cancer cell models. The material is supplied as a high-purity solid for biochemical and cellular assays.
  • Potent class I HDAC inhibitor with low-nanomolar activity.
  • Demonstrated in vitro and in vivo antitumor activity.
  • Induces apoptosis and histone acetylation in cell studies.
  • Supplied as a dihydrochloride salt suitable for handling and formulation.
  • High purity (≥99.0%) appropriate for research assays.
  • Storage: 4°C sealed; in solution keep at -80°C for long-term storage.
Catalog No. 50-002-62003