Learn More
Medchemexpress LLC HDAC-IN-4 | 1252003-13-6 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12876310MM/1ML
Encompass_Preferred
HDAC-IN-4 is a selective inhibitor of HDAC6 and HDAC10, with pIC50 values of 7.2 and 6.8 respectively in a BRET assay. This compound demonstrates antitumoral activity and is noted to be more potent against HDAC6 than Tubastatin A.
  • Selective inhibition of HDAC6 and HDAC10
  • Exhibits antitumoral activity
  • More potent against HDAC6 compared to Tubastatin A
  • Inhibits HDAC1, HDAC2, HDAC3, HDAC8, and HDAC10 in FRET assay
  • For research use only
Catalog No. 50-003-39378